4-[(Benzyloxy)methyl]-6-chloropyrimidine 95+%

Lieferant: Apollo Scientific

OR70223-250MG
APOSOR70223-250MGEA 173 EUR
APOSOR70223-250MG
4-[(Benzyloxy)methyl]-6-chloropyrimidine 95+%
4-[(Benzyloxy)methyl]-6-chloropyrimidine
Formel: C₁₂H₁₁ClN₂O
Molecular Weight: 234,682 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09038781
CAS-Nummer: 914802-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR