3-(Dimethylamino)-1-(2,4,5-trichlorophenyl)prop-2-en-1-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(E)-3-(Dimethylamino)-1-(2,4,5-trichlorophenyl)prop-2-en-1-one
OR70231-250MG
APOSOR70231-250MGEA
219
EUR
APOSOR70231-250MG
3-(Dimethylamino)-1-(2,4,5-trichlorophenyl)prop-2-en-1-one
3-(Dimethylamino)-1-(2,4,5-trichlorophenyl)prop-2-en-1-one
Formel:
C₁₁H₁₀Cl₃NO Molecular Weight: 278,56 g/mol |
MDL:
MFCD32862647 CAS-Nummer: 2645406-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...