N,N-Dimethylpyridine-3-sulphonamide 98%

Lieferant: Apollo Scientific
Achtung

OR7025-1G
APOSOR7025-1GEA 176 EUR
APOSOR7025-1G
N,N-Dimethylpyridine-3-sulphonamide 98%
N,N-Dimethylpyridine-3-sulphonamide
Formel: C₇H₁₀N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD07368248
CAS-Nummer: 4810-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR