3-Amino-4,6-dimethylpyridin 97%

Lieferant: Apollo Scientific

Synonyme: 5A-DMP, 4,6-Dimethylpyridin-3-amine

OR7030-250MG
APOSOR7030-250MGEA 21.9 EUR
APOSOR7030-250MG
3-Amino-4,6-dimethylpyridin 97%
3-Amino-4,6-dimethylpyridine
Formel: C₇H₁₀N₂
Molecular Weight: 122,1677 g/mol
MDL: MFCD07439969
CAS-Nummer: 1193-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR