2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile, 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile

OR7031-1G
APOSOR7031-1GEA 21.9 EUR
APOSOR7031-1G
2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
Formel: C₉H₁₀N₂S
Molecular Weight: 178,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00128278
CAS-Nummer: 4651-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR