2-((2-Methylbut-3-yn-2-yl)oxy)tetrahydro-2H-pyran 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Methylbut-3-yn-2-yloxy)oxane

OR70332-1G OR70332-5G OR70332-25G
APOSOR70332-1GEA 21.9 EUR
APOSOR70332-1G APOSOR70332-5G APOSOR70332-25G
2-((2-Methylbut-3-yn-2-yl)oxy)tetrahydro-2H-pyran 97%
2-((2-Methylbut-3-yn-2-yl)oxy)tetrahydro-2H-pyran
Formel: C₁₀H₁₆O₂
Molecular Weight: 168.23 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00090526
CAS-Nummer: 27943-46-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR