Benzoimidazol-1-ylacetic acid 95%

Lieferant: Apollo Scientific
Achtung

OR7034-1G
APOSOR7034-1GEA 0 EUR
APOSOR7034-1G
Benzoimidazol-1-ylacetic acid 95%
Benzoimidazol-1-ylacetic acid
Formel: C₉H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00956879
CAS-Nummer: 40332-16-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR