1-(3-Aminoprop-1-yl)-1H-benzimidazole hydrochloride

Lieferant: Apollo Scientific
Achtung

OR7038-1G
APOSOR7038-1GEA 0 EUR
APOSOR7038-1G
1-(3-Aminoprop-1-yl)-1H-benzimidazole hydrochloride
1-(3-Aminoprop-1-yl)-1H-benzimidazole hydrochloride
Formel: C₁₀H₁₃N₃·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD04148859
CAS-Nummer: 1048649-78-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR