Sodium (2-amino-4-bromo-5-methylphenyl)(oxo)acetate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-5-methylisatinic acid sodium salt, Sodium 2-amino-4-bromo-5-methylphenylglyoxylate
OR7043-1G
APOSOR7043-1GEA
267
EUR
APOSOR7043-1G
Sodium (2-amino-4-bromo-5-methylphenyl)(oxo)acetate
Sodium (2-amino-4-bromo-5-methylphenyl)(oxo)acetate
Formel:
C₉H₇BrNO₃·Na Molecular Weight: 280,051 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093056 CAS-Nummer: 1171690-78-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...