3-(4-Bromophenyl)cyclobutan-1-one
Lieferant: Apollo Scientific
Synonyme:
1-(4-Bromophenyl)-3-oxocyclobutane, 3-(4-Bromophenyl)cyclobutanone
OR7045-1G
APOSOR7045-1GEA
232
EUR
APOSOR7045-1G
3-(4-Bromophenyl)cyclobutan-1-one
3-(4-Bromophenyl)cyclobutan-1-one
Formel:
C₁₀H₉BrO Molecular Weight: 225,082 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038979 CAS-Nummer: 254892-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...