1-Cyclopentylazetidine-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Carboxy-1-cyclopentylazetidine, 1-Cyclopentylazetane-2-carboxylic acid
OR7049-1G
APOSOR7049-1GEA
395
EUR
APOSOR7049-1G
1-Cyclopentylazetidine-2-carboxylic acid
1-Cyclopentylazetidine-2-carboxylic acid
Formel:
C₉H₁₅NO₂ Molecular Weight: 169.221 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038981 CAS-Nummer: 1042413-79-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...