1-Cyclopropylethylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR7051-10G OR7051-1G OR7051-5G
APOSOR7051-10GEA 575 EUR
APOSOR7051-10G APOSOR7051-1G APOSOR7051-5G
1-Cyclopropylethylamine hydrochloride
1-Cyclopropylethylamine hydrochloride
Formel: C₅H₁₂ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD06685893
CAS-Nummer: 42390-64-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR