4-(2-Bromoethyl)-3,5-dimethyl-1H-pyrazole
Lieferant: Apollo Scientific
|
Gefahr
|
OR7065-100MG
APOSOR7065-100MGEA
43.5
EUR
APOSOR7065-100MG
4-(2-Bromoethyl)-3,5-dimethyl-1H-pyrazole
4-(2-Bromoethyl)-3,5-dimethyl-1H-pyrazole
Formel:
C₇H₁₁BrN₂ Schmelzpunkt: 118…120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00480987 CAS-Nummer: 83467-28-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...