5-Propyl-1,3-cyclohexandion
Lieferant: Apollo Scientific
OR7071-1G
APOSOR7071-1GEA
132
EUR
APOSOR7071-1G
5-Propyl-1,3-cyclohexandion
5-Propyl-1,3-cyclohexanedione
Formel:
C₉H₁₄O₂ |
MDL:
MFCD00129456 CAS-Nummer: 57641-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...