(3,5-Dichloro-4-ethoxyphenyl)methanol 97%

Lieferant: Apollo Scientific

OR71008-500MG
APOSOR71008-500MGEA 736 EUR
APOSOR71008-500MG
(3,5-Dichloro-4-ethoxyphenyl)methanol 97%
(3,5-Dichloro-4-ethoxyphenyl)methanol
Formel: C₉H₁₀Cl₂O₂
Molecular Weight: 221.081 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03862359
CAS-Nummer: 664319-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR