[2-(1H-1,2,4-Triazol-1-yl)ethyl]amine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR7106-50MG
APOSOR7106-50MGEA 0 EUR
APOSOR7106-50MG
[2-(1H-1,2,4-Triazol-1-yl)ethyl]amine hydrochloride 95%
[2-(1H-1,2,4-Triazol-1-yl)ethyl]amine hydrochloride
Formel: C₄H₉ClN₄
Lagertemperatur: Raumtemperatur
MDL: MFCD06656249
CAS-Nummer: 51444-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR