Ethyl 3-cyano-1H-1,2,4-triazole-5-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

OR71096-250MG
APOSOR71096-250MGEA 473 EUR
APOSOR71096-250MG
Ethyl 3-cyano-1H-1,2,4-triazole-5-carboxylate 95%
Ethyl 3-cyano-1H-1,2,4-triazole-5-carboxylate
Formel: C₆H₆N₄O₂
Molecular Weight: 166,14 g/mol
MDL: MFCD14155905
CAS-Nummer: 853907-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR