(4-Methoxymethylphenyl)oxoacetic acid

Lieferant: Apollo Scientific
Achtung

OR71103-1G
APOSOR71103-1GEA 629 EUR
APOSOR71103-1G
(4-Methoxymethylphenyl)oxoacetic acid
(4-Methoxymethylphenyl)oxoacetic acid
Formel: C₁₁H₁₂O₄
Molecular Weight: 208,21 g/mol
MDL: MFCD23143081
CAS-Nummer: 1443339-17-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR