N-Ethylpyridine-3-sulphonamide 96%

Lieferant: Apollo Scientific

Synonyme: 3-Ethylsulphamoylpyridine

OR7151-1G
APOSOR7151-1GEA 176 EUR
APOSOR7151-1G
N-Ethylpyridine-3-sulphonamide 96%
N-Ethylpyridine-3-sulphonamide
Formel: C₇H₁₀N₂O₂S
Molecular Weight: 186,232 g/mol
Schmelzpunkt: 89…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027298
CAS-Nummer: 4810-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR