2-Benzyloxy-5-chlorphenylboronsäure

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2-(Benzyloxy)-5-chlorophenyl)boronic acid

OR7153-5G
APOSOR7153-5GEA 157 EUR
APOSOR7153-5G
2-Benzyloxy-5-chlorphenylboronsäure
2-Benzyloxy-5-chlorphenylboronsäure
Formel: C₁₃H₁₂BClO₃
Molecular Weight: 262.4966 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039000
CAS-Nummer: 612832-83-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR