4-Benzyloxy-3,5-dichlorphenylboronsäure
Lieferant: Apollo Scientific
|
Gefahr
|
OR7155-1G
OR7155-250MG
OR7155-5G
APOSOR7155-1GEA
128
EUR
APOSOR7155-1G
APOSOR7155-250MG
APOSOR7155-5G
4-Benzyloxy-3,5-dichlorphenylboronsäure
4-Benzyloxy-3,5-dichlorphenylboronsäure
Formel:
C₁₃H₁₁BCl₂O₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD04039002 CAS-Nummer: 1003298-85-8 UN: 1759 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...