4-Benzyloxy-3,5-dichlorphenylboronsäure

Lieferant: Apollo Scientific
Gefahr

OR7155-1G OR7155-250MG OR7155-5G
APOSOR7155-1GEA 128 EUR
APOSOR7155-1G APOSOR7155-250MG APOSOR7155-5G
4-Benzyloxy-3,5-dichlorphenylboronsäure
4-Benzyloxy-3,5-dichlorphenylboronsäure
Formel: C₁₃H₁₁BCl₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD04039002
CAS-Nummer: 1003298-85-8
UN: 1759
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR