3-(tert-Butoxymethyl)phenylboronic acid 98%

Lieferant: Apollo Scientific
Achtung

OR7174-1G OR7174-5G
APOSOR7174-1GEA 29 EUR
APOSOR7174-1G APOSOR7174-5G
3-(tert-Butoxymethyl)phenylboronic acid 98%
3-(tert-Butoxymethyl)phenylboronic acid
Formel: C₁₁H₁₇BO₃
Lagertemperatur: Kühlschrank
MDL: MFCD03788411
CAS-Nummer: 858364-78-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR