4-(tert-Butoxymethyl)phenylboronic acid 98%

Lieferant: Apollo Scientific
Achtung

OR7176-1G OR7176-250MG OR7176-5G
APOSOR7176-1GEA 24.8 EUR
APOSOR7176-1G APOSOR7176-250MG APOSOR7176-5G
4-(tert-Butoxymethyl)phenylboronic acid 98%
4-(tert-Butoxymethyl)phenylboronic acid
Formel: C₁₁H₁₇BO₃
Lagertemperatur: Kühlschrank
MDL: MFCD03701685
CAS-Nummer: 1024017-53-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR