trans-N-Boc-1,4-cyclohexanediamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
trans-4-(Boc-amino)cyclohexylamine, trans-tert-Butyl (4-aminocyclohexyl)carbamate
OR7177-1G
OR7177-5G
OR7177-25G
OR7177-100G
APOSOR7177-1GEA
16.1
EUR
APOSOR7177-1G
APOSOR7177-5G
APOSOR7177-25G
APOSOR7177-100G
trans-N-Boc-1,4-cyclohexanediamine
trans-N-Boc-1,4-cyclohexanediamine
Formel:
C₁₁H₂₂N₂O₂ Schmelzpunkt: 74,5…75.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001719 CAS-Nummer: 177906-48-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...