2-Chlor-5-pyridinboronsäurepinakolester 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(6-Chloro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Chloropyridine-5-boronic acid Pinacol ester, 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
OR7183-5G
APOSOR7183-5GEA
39.3
EUR
APOSOR7183-5G
2-Chlor-5-pyridinboronsäurepinakolester 97%
2-Chlor-5-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₅BClNO₂ Molecular Weight: 239.51 g/mol Schmelzpunkt: 86…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03412793 CAS-Nummer: 444120-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...