2-Cyanobenzeneboronic acid-1,3-propanediol ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(1,3,2-Dioxaborinan-2-yl)benzonitrile, 2-Cyanobenzeneboronic acid 1,3-Propanediol ester, 2-Cyanophenylboronic acid 1,3-Propanediol ester
OR7188-1G
APOSOR7188-1GEA
21.9
EUR
APOSOR7188-1G
2-Cyanobenzeneboronic acid-1,3-propanediol ester
2-Cyanobenzeneboronic acid-1,3-propanediol ester
Formel:
C₁₀H₁₀BNO₂ Molecular Weight: 187,00 g/mol Schmelzpunkt: 42…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039012 CAS-Nummer: 172732-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...