2-(Diethylcarbamoyl)phenylboronsäure 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2-(Diethylcarbamoyl)phenyl)boronic acid

OR7197-1G
APOSOR7197-1GEA 227 EUR
APOSOR7197-1G
2-(Diethylcarbamoyl)phenylboronsäure 95%
2-(Diethylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₃
Molecular Weight: 221,0600 g/mol
Schmelzpunkt: 64…70 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01318988
CAS-Nummer: 129112-21-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR