2-(2-tert-Butyldimethylsilyloxyethyl)bromobenzene 98%
Lieferant: Apollo Scientific
OR7200-1G
APOSOR7200-1GEA
176
EUR
APOSOR7200-1G
2-(2-tert-Butyldimethylsilyloxyethyl)bromobenzene 98%
2-(2-tert-Butyldimethylsilyloxyethyl)bromobenzene
Formel:
C₁₄H₂₃BrOSi Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258648 CAS-Nummer: 181021-20-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...