1,1-Diiodoferrocene
Lieferant: Apollo Scientific
|
Achtung
|
OR72053-250MG
APOSOR72053-250MGEA
87
EUR
APOSOR72053-250MG
1,1-Diiodoferrocene
1,1-Diiodoferrocene
Formel:
C₁₀H₈FeI₂ Lagertemperatur: Kühlschrank |
CAS-Nummer:
12145-93-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...