3-Formyl-4-nitrobenzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR72159-1G
APOSOR72159-1GEA
783
EUR
APOSOR72159-1G
3-Formyl-4-nitrobenzoic acid 95%
3-Formyl-4-nitrobenzoic acid
Formel:
C₈H₅NO₅ |
MDL:
MFCD22200133 CAS-Nummer: 1092931-93-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...