4-Formylbenzeneboronic acid, propane-1,3-diol cyclic ester
Lieferant: Apollo Scientific
|
Achtung
|
OR7217-1G
OR7217-500MG
APOSOR7217-1GEA
49.3
EUR
APOSOR7217-1G
APOSOR7217-500MG
4-Formylbenzeneboronic acid, propane-1,3-diol cyclic ester
4-Formylbenzeneboronic acid, propane-1,3-diol cyclic ester
Formel:
C₁₀H₁₁BO₃ Molecular Weight: 190.004 g/mol Schmelzpunkt: 72…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039026 CAS-Nummer: 4463-41-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...