4-Formylbenzeneboronic acid, neopentyl glycol ester 98%

Lieferant: Apollo Scientific
Achtung

OR7219-5G
APOSOR7219-5GEA 34.4 EUR
APOSOR7219-5G
4-Formylbenzeneboronic acid, neopentyl glycol ester 98%
4-Formylbenzeneboronic acid, neopentyl glycol ester
Formel: C₁₂H₁₅BO₃
Siedepunkt: 350 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06796332
CAS-Nummer: 128376-65-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR