2-Amino-3,5-dinitro-6-picolin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dinitro-6-methyl-2-pyridylamin, 2-Amino-3,5-dinitro-6-methylpyridin

OR7220-1G
APOSOR7220-1GEA 176 EUR
APOSOR7220-1G
2-Amino-3,5-dinitro-6-picolin 97%
2-Amino-3,5-dinitro-6-picolin
Formel: C₆H₆N₄O₄
Molecular Weight: 198,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00091867
CAS-Nummer: 25864-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR