4'-Methoxybiphenyl-4-ylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Methoxyphenyl)benzeneboronic acid, 4-(4-Methoxyphenyl)phenylboronic acid, (4'-Methoxy-[1,1'-biphenyl]-4-yl)boronic acid

OR7229-1G
APOSOR7229-1GEA 163 EUR
APOSOR7229-1G
4'-Methoxybiphenyl-4-ylboronsäure
4'-Methoxybiphenyl-4-ylboronsäure
Formel: C₁₃H₁₃BO₃
Molecular Weight: 228,0515 g/mol
Schmelzpunkt: 209…211 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04039030
CAS-Nummer: 156642-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR