3-(Methoxymethyl)phenylboronic acid

Lieferant: Apollo Scientific
Achtung

OR7232-1G OR7232-250MG OR7232-5G
APOSOR7232-1GEA 150 EUR
APOSOR7232-1G APOSOR7232-250MG APOSOR7232-5G
3-(Methoxymethyl)phenylboronic acid
3-(Methoxymethyl)phenylboronic acid
Formel: C₈H₁₁BO₃
Lagertemperatur: Kühlschrank
MDL: MFCD03412069
CAS-Nummer: 142273-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR