6-Methoxy-2-naphthalinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methoxy-2-naphthylboronic acid, 6-Methoxy-2-naphthylboronic Acid

OR7234-25G OR7234-5G OR7234-1G OR7234-10G
APOSOR7234-25GEA 101 EUR
APOSOR7234-25G APOSOR7234-5G APOSOR7234-1G APOSOR7234-10G
6-Methoxy-2-naphthalinboronsäure
6-Methoxy-2-naphthaleneboronic acid
Formel: C₁₁H₁₁BO₃
Molecular Weight: 202.02 g/mol
Schmelzpunkt: 311 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03093087
CAS-Nummer: 156641-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR