2-[(2-Methylphenoxy)methyl]benzeneboronic acid

Lieferant: Apollo Scientific
Achtung

OR7239-100MG
APOSOR7239-100MGEA 43.7 EUR
APOSOR7239-100MG
2-[(2-Methylphenoxy)methyl]benzeneboronic acid
2-[(2-Methylphenoxy)methyl]benzeneboronic acid
Formel: C₁₄H₁₅BO₃
Molecular Weight: 242,078 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04039032
CAS-Nummer: 170924-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR