4-(Morpholino)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Morpholinophenyl)boronic acid

OR7242-1G OR7242-250MG OR7242-5G
APOSOR7242-1GEA 24.8 EUR
APOSOR7242-1G APOSOR7242-250MG APOSOR7242-5G
4-(Morpholino)phenylboronsäure
4-(Morpholino)phenylboronsäure
Formel: C₁₀H₁₄BNO₃
Schmelzpunkt: 126…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095169
CAS-Nummer: 186498-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR