3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)acrylonitrile

Lieferant: Apollo Scientific
Gefahr

OR7250T-100MG OR7250T-1G OR7250T-500MG
APOSOR7250T-100MGEA 0 EUR
APOSOR7250T-100MG APOSOR7250T-1G APOSOR7250T-500MG
3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)acrylonitrile
3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)acrylonitrile

A reference standard for the drug Trimethoprim.

Formel: C₁₉H₂₀N₂O₃
Schmelzpunkt: 130…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00269911
CAS-Nummer: 30078-48-9
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR