3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)acrylonitrile
Lieferant: Apollo Scientific
|
Gefahr
|
OR7250T-100MG
OR7250T-1G
OR7250T-500MG
APOSOR7250T-100MGEA
0
EUR
APOSOR7250T-100MG
APOSOR7250T-1G
APOSOR7250T-500MG
3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)acrylonitrile
3-(Phenylamino)-2-(3,4,5-trimethoxybenzyl)acrylonitrile
A reference standard for the drug Trimethoprim.
Formel:
C₁₉H₂₀N₂O₃ Schmelzpunkt: 130…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269911 CAS-Nummer: 30078-48-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...