4-Amino-3,5-dichlorbenzonitril 98%

Lieferant: Apollo Scientific
Achtung

OR7252-5G
APOSOR7252-5GEA 108 EUR
APOSOR7252-5G
4-Amino-3,5-dichlorbenzonitril 98%
4-Amino-3,5-dichlorbenzonitril
Formel: C₇H₄Cl₂N₂
Molecular Weight: 187,03 g/mol
Schmelzpunkt: 116…120 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00017341
CAS-Nummer: 78473-00-4
EINECS: 278-914-6
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR