3'-Amino-[1,1'-biphenyl]-3-carboxylic acid hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3'-Amino-3-carboxybiphenyl hydrochloride, 3-(3-Aminophenyl)benzoic acid hydrochloride

OR7317-100MG
APOSOR7317-100MGEA 21.9 EUR
APOSOR7317-100MG
3'-Amino-[1,1'-biphenyl]-3-carboxylic acid hydrochloride
3'-Amino-[1,1'-biphenyl]-3-carboxylic acid hydrochloride
Formel: C₁₃H₁₁NO₂·ClH
Molecular Weight: 249,693 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08456987
CAS-Nummer: 1170469-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR