2'-(Benzyloxy)-[1,1'-biphenyl]-2-amine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-2'-(benzyloxy)biphenyl hydrochloride, 2-[2-(Benzyloxy)phenyl]aniline hydrochloride

OR7321-250MG
APOSOR7321-250MGEA 21.9 EUR
APOSOR7321-250MG
2'-(Benzyloxy)-[1,1'-biphenyl]-2-amine hydrochloride
2'-(Benzyloxy)-[1,1'-biphenyl]-2-amine hydrochloride
Formel: C₁₉H₁₇NO·ClH
Molecular Weight: 311,805 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07366472
CAS-Nummer: 1170524-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR