4'-(Benzyloxy)-[1,1'-biphenyl]-2-amine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[4-(Benzyloxy)phenyl]aniline, 2-Amino-4'-(benzyloxy)biphenyl

OR7323-250MG
APOSOR7323-250MGEA 21.9 EUR
APOSOR7323-250MG
4'-(Benzyloxy)-[1,1'-biphenyl]-2-amine 98%
4'-(Benzyloxy)-[1,1'-biphenyl]-2-amine
Formel: C₁₉H₁₇NO
Molecular Weight: 275,344 g/mol
Schmelzpunkt: 116…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039047
CAS-Nummer: 400746-75-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR