2-Amino-3'-chlorobiphenyl hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Chlorophenyl)aniline hydrochloride, 3'-Chloro-[1,1'-biphenyl]-2-amine hydrochloride

OR7337-50MG
APOSOR7337-50MGEA 36.4 EUR
APOSOR7337-50MG
2-Amino-3'-chlorobiphenyl hydrochloride
2-Amino-3'-chlorobiphenyl hydrochloride
Formel: C₁₂H₁₀ClN·ClH
Molecular Weight: 240,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739421
CAS-Nummer: 1172032-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR