2-Amino-4'-chlorobiphenyl

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Chloro-[1,1'-biphenyl]-2-amine

OR7338-10MG
APOSOR7338-10MGEA 21.9 EUR
APOSOR7338-10MG
2-Amino-4'-chlorobiphenyl
2-Amino-4'-chlorobiphenyl
Formel: C₁₂H₁₀ClN
Molecular Weight: 203,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424659
CAS-Nummer: 1204-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR