1,1-Diphenylethylene oxide 97%

Lieferant: Apollo Scientific
Gefahr

OR7342-10G OR7342-1G OR7342-5G OR7342-25G
APOSOR7342-10GEA 0 EUR
APOSOR7342-10G APOSOR7342-1G APOSOR7342-5G APOSOR7342-25G
1,1-Diphenylethylene oxide 97%
1,1-Diphenylethylene oxide
Formel: C₁₄H₁₂O
Schmelzpunkt: 54…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040725
CAS-Nummer: 882-59-7
UN: 1325
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR