1-O-Acetyl-2-deoxy-3,5-di-O-benzoylribofuranose

Lieferant: Apollo Scientific
Achtung

OR7350T-250MG OR7350T-50MG
APOSOR7350T-250MGEA 865 EUR
APOSOR7350T-250MG APOSOR7350T-50MG
1-O-Acetyl-2-deoxy-3,5-di-O-benzoylribofuranose
1-O-Acetyl-2-deoxy-3,5-di-O-benzoylribofuranose
Formel: C₂₁H₂₀O₇
Lagertemperatur: Raumtemperatur
MDL: MFCD00269868
CAS-Nummer: 51255-12-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR