Dansylamidoethyl methanethiosulphonate

Lieferant: Apollo Scientific
Achtung

Synonyme: S-Methyl 2-({[5-(Dimethylamino)naphth-1-yl]sulphonyl}amino)ethanesulphonothioate, 2-{[5-(Dimethylamino)naphth-1-yl]sulphonamido}ethyl methanethiosulphonate, MTS-Dansyl

OR7352T-10MG
APOSOR7352T-10MGEA 259 EUR
APOSOR7352T-10MG
Dansylamidoethyl methanethiosulphonate
Dansylamidoethyl methanethiosulphonate

A florescent MTS reagent that reacts specifically & rapidlywith thiols.

Formel: C₁₅H₂₀N₂O₄S₃
Molecular Weight: 388.525 g/mol
Schmelzpunkt: 94…97 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD08445467
CAS-Nummer: 355115-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR